Learn More
Meso Scale Discovery S-PLEX SARS-CoV-2 Spike KitQP
SDP
Catalog No. 5000078816
Encompass
The S-PLEX SARS-CoV-2 Spike Kit from Meso Scale Diagnostics is an electrochemiluminescent immunoassay that quantitatively measures antibodies specific to the SARS-CoV-2 spike protein This multiplex kit includes a 96-well plate coated with the SARS-CoV-2 spike protein receptor binding domain along with calibrators and controls The assay has a 3 5 hour turnaround time and requires a minimal sample volume of 25 L of serum or plasma This kit allows for high-throughput quantitative antibody testing to aid in COVID-19 vaccine development and immune response monitoring
Catalog No. 50-000-78816 Supplier Meso Scale Discovery Supplier No. K150ADJS22
Add to Cart
Add to Cart
Request a Quote
The S-PLEX SARS-CoV-2 Spike Kit from Meso Scale Diagnostics is an electrochemiluminescent immunoassay that quantitatively measures antibodies specific to the SARS-CoV-2 spike protein This multiplex kit includes a 96-well plate coated with the SARS-CoV-2 spike protein receptor binding domain along with calibrators and controls The assay has a 3 5 hour turnaround time and requires a minimal sample volume of 25 L of serum or plasma This kit allows for high-throughput quantitative antibody testing to aid in COVID-19 vaccine development and immune response monitoring