Learn More
Carbosynth LLC. Mouse monoclonal Streptococcus pneumoniae antibody
SDP
Supplier:  Carbosynth LLC. 10S27C0.2MG
Encompass_Preferred
Streptococcus pneumoniae, or pneumococcus, is Gram-positive, alpha-hemolytic, bile-soluble aerotolerant, anaerobic member of the genus Streptococcus. S. pneumoniae is the most common cause of bacterial meningitis in adults, children, and dogs, and is one of the top two isolates found in ear infection, otitis media. The genome of S. pneumoniae is a closed, circular DNA structure that contains between 2.0 and 2.1 million basepairs.
Catalog No. 50-125-2799