Learn More
Medchemexpress LLC HY-139400 5mg , (S)-Pociredir CAS:2490676-18-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1394005MG
Encompass_Preferred
HY-139400 5mg (S)-Pociredir CAS: 2490676-18-9 (S)-Pociredir ((S)-FTX-6058) is a potent and orally active inhibitor of Embryonic Ectoderm Development (EED). (S)-Pociredir can induce HbF protein expression in cell and murine models. (S)-Pociredir can be used for the research of select hemoglobinopathies, including sickle cell disease and β-thalassemia. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0118