Learn More
Medchemexpress LLC S.pombe lumazine synthase-IN-1 | 331726-35-3 | 98.0% | 319.27 | 10 MM * 1 ML
SDP
Catalog No. 5000405123
Encompass_Preferred
S.pombe lumazine synthase-IN-1 is an inhibitor of lumazine synthases, demonstrating Ki values of 243 μM for *Schizosaccharomyces pombe* and 9.6 μM for *Mycobacterium tuberculosis* lumazine synthases. It is intended for research use only.
  • Inhibitor of lumazine synthases.
  • Specific Ki values for *Schizosaccharomyces pombe* (243 μM) and *Mycobacterium tuberculosis* (9.6 μM).
  • Class II compound with OCH3 groups showing slightly better potencies compared to counterparts with OH groups.
  • Provided with Data Sheet, COA, SDS, and Handling Instructions.
  • Available in various forms including solid and solution in DMSO.
  • Can be stored as powder at -20°C for 3 years or 4°C for 2 years, and in solvent at -80°C for 6 months or -20°C for 1 month.
  • Related to anti-infection, bacterial infection, fungal infection, and *Mycobacterium tuberculosis* infection research.
Catalog No. 50-004-05123 Supplier Medchemexpress LLC Supplier No. HY4468810MM/1ML
S.pombe lumazine synthase-IN-1 is an inhibitor of lumazine synthases, demonstrating Ki values of 243 μM for *Schizosaccharomyces pombe* and 9.6 μM for *Mycobacterium tuberculosis* lumazine synthases. It is intended for research use only.
  • Inhibitor of lumazine synthases.
  • Specific Ki values for *Schizosaccharomyces pombe* (243 μM) and *Mycobacterium tuberculosis* (9.6 μM).
  • Class II compound with OCH3 groups showing slightly better potencies compared to counterparts with OH groups.
  • Provided with Data Sheet, COA, SDS, and Handling Instructions.
  • Available in various forms including solid and solution in DMSO.
  • Can be stored as powder at -20°C for 3 years or 4°C for 2 years, and in solvent at -80°C for 6 months or -20°C for 1 month.
  • Related to anti-infection, bacterial infection, fungal infection, and *Mycobacterium tuberculosis* infection research.