Learn More
Microsolv Technology Corporation S Pp Screw 700Ul Lcm
Catalog No. NC1513550
Encompass_Preferred
Vials, Screw Top, Plastic. Clear, 700ul MicroVial, LCMS Compatible, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial or storage bottle. AQ Brand. 100/PK.
Catalog No. NC1513550 Supplier Microsolv Technology Corporation Supplier No. 9572SMS
Vials, Screw Top, Plastic. Clear, 700ul MicroVial, LCMS Compatible, 9-425mm threads, 12x32mm outer dimensions. For use as an autosampler vial or storage bottle. AQ Brand. 100/PK.