Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-praziquantel-d11 | 00-00-0 | 98.2% | 323.47 g/mol | C19H13D11N2O2 | 5 MG
SDP

Catalog No. 5000264812
Encompass_Preferred

(S)-Praziquantel-d11 is the deuterium-labeled S-enantiomer of praziquantel supplied as a research standard and analytical reference. It is intended for laboratory use as an internal standard or tracer in mass spectrometry and other analytical workflows; not for human or clinical use.

  • Deuterium-labeled S-enantiomer for use as an internal standard.
  • High purity: 98.2%.
  • Chemical formula: C19H13D11N2O2.
  • Molecular weight: 323.47 g/mol.
  • Supplied as a small-quantity powder suitable for analytical applications.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-64812 Supplier Medchemexpress LLC Supplier No. HY126057AS5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S)-Praziquantel-d11 is the deuterium-labeled S-enantiomer of praziquantel supplied as a research standard and analytical reference. It is intended for laboratory use as an internal standard or tracer in mass spectrometry and other analytical workflows; not for human or clinical use.

  • Deuterium-labeled S-enantiomer for use as an internal standard.
  • High purity: 98.2%.
  • Chemical formula: C19H13D11N2O2.
  • Molecular weight: 323.47 g/mol.
  • Supplied as a small-quantity powder suitable for analytical applications.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.