Learn More
Medchemexpress LLC (S)-Pro-xylane | 868156-46-1 | MFCD32701904 | 98.0% | 192.21 g/mol | C8H16O5 | 100 MG
SDP
Catalog No. 5000242981
Encompass_Preferred
(S)-Pro-xylane ((S)-Hydroxypropyl tetrahydropyrantriol) is a bioactive C-glycoside used in cosmetic and dermatological research to stimulate glycosaminoglycan biosynthesis and support skin extracellular matrix health. Supplied as a white to off-white solid, it is intended for formulation, in vitro, and topical delivery studies.
  • Stimulates biosynthesis of glycosaminoglycans (GAGs) in fibroblasts.
  • Enhances structural stability of the skin extracellular matrix and improves elasticity.
  • Promotes collagen and hyaluronic acid synthesis in the dermis.
  • Suitable for topical formulation and transdermal absorption studies.
  • High purity solid appropriate for research applications.
  • Soluble in DMSO and water with ultrasonic assistance for stock preparation.
Catalog No. 50-002-42981 Supplier Medchemexpress LLC Supplier No. HY108036A100MG
(S)-Pro-xylane ((S)-Hydroxypropyl tetrahydropyrantriol) is a bioactive C-glycoside used in cosmetic and dermatological research to stimulate glycosaminoglycan biosynthesis and support skin extracellular matrix health. Supplied as a white to off-white solid, it is intended for formulation, in vitro, and topical delivery studies.
  • Stimulates biosynthesis of glycosaminoglycans (GAGs) in fibroblasts.
  • Enhances structural stability of the skin extracellular matrix and improves elasticity.
  • Promotes collagen and hyaluronic acid synthesis in the dermis.
  • Suitable for topical formulation and transdermal absorption studies.
  • High purity solid appropriate for research applications.
  • Soluble in DMSO and water with ultrasonic assistance for stock preparation.