Learn More
Medchemexpress LLC (S)-pro-xylane | 868156-46-1 | MFCD32701904 | 98.0% | 192.21 g/mol | C8H16O5 | 50 MG
SDP
Catalog No. 5000242983
Encompass_Preferred
(S)-Pro-xylane is the S-enantiomer of pro-xylane (hydroxypropyl tetrahydropyrantriol), a bioactive C-glycoside used in research on the skin extracellular matrix and cosmetic active ingredient development. It stimulates glycosaminoglycan biosynthesis, promotes collagen and hyaluronic acid production, and is supplied as a research-grade solid with supporting documentation available.
  • Stimulates biosynthesis of glycosaminoglycans (GAGs) in fibroblasts.
  • Promotes collagen and hyaluronic acid synthesis in dermal tissue.
  • Improves skin elasticity and hydration in topical studies.
  • Eco-friendly and biodegradable profile.
  • Provided with purity data, COA, and SDS for research use.
  • For research use only; not for human therapeutic use.
Catalog No. 50-002-42983 Supplier Medchemexpress LLC Supplier No. HY108036A50MG
(S)-Pro-xylane is the S-enantiomer of pro-xylane (hydroxypropyl tetrahydropyrantriol), a bioactive C-glycoside used in research on the skin extracellular matrix and cosmetic active ingredient development. It stimulates glycosaminoglycan biosynthesis, promotes collagen and hyaluronic acid production, and is supplied as a research-grade solid with supporting documentation available.
  • Stimulates biosynthesis of glycosaminoglycans (GAGs) in fibroblasts.
  • Promotes collagen and hyaluronic acid synthesis in dermal tissue.
  • Improves skin elasticity and hydration in topical studies.
  • Eco-friendly and biodegradable profile.
  • Provided with purity data, COA, and SDS for research use.
  • For research use only; not for human therapeutic use.