Learn More
Medchemexpress LLC S-Propargylcysteine | 3262-64-4 | 159.21 | 250 MG
SDP
Catalog No. 5000450060
Encompass_Preferred
S-Propargylcysteine (SPRC) is a structural analog of S-allyl cysteine (SAC) and functions as a slow H2S-releasing compound. It reduces Ca2+ accumulation and inflammatory cytokines, inhibits STAT3, and elevates p53 and Bax. SPRC exhibits anti-inflammatory activity and offers protection against acute pancreatitis in mice. Additionally, it possesses cardioprotective and neuroprotective properties.
  • Reduces Ca2+ accumulation and inflammatory cytokines
  • Inhibits STAT3
  • Elevates p53 and Bax
  • Exhibits anti-inflammatory activity
  • Protects against acute pancreatitis in mice
  • Possesses cardioprotective properties
  • Possesses neuroprotective properties
Catalog No. 50-004-50060 Supplier Medchemexpress LLC Supplier No. 250MGHYW001538
S-Propargylcysteine (SPRC) is a structural analog of S-allyl cysteine (SAC) and functions as a slow H2S-releasing compound. It reduces Ca2+ accumulation and inflammatory cytokines, inhibits STAT3, and elevates p53 and Bax. SPRC exhibits anti-inflammatory activity and offers protection against acute pancreatitis in mice. Additionally, it possesses cardioprotective and neuroprotective properties.
  • Reduces Ca2+ accumulation and inflammatory cytokines
  • Inhibits STAT3
  • Elevates p53 and Bax
  • Exhibits anti-inflammatory activity
  • Protects against acute pancreatitis in mice
  • Possesses cardioprotective properties
  • Possesses neuroprotective properties