Learn More
Selleck Chemical LLC S-Propranolol hydrochloride
SDP
Catalog No. 5000004063
Encompass_Preferred
(S)-(-)-Propranolol hydrochloride the active enantiomer of propranolol is a -Adrenoceptor ( -Adrenergic Receptor) antagonist with log Kd values of -8 16 -9 08 and -6 93 for 1 2 and 3 respectively
Catalog No. 50-000-04063 Supplier Selleck Chemical LLC Supplier No. S59811G
(S)-(-)-Propranolol hydrochloride the active enantiomer of propranolol is a -Adrenoceptor ( -Adrenergic Receptor) antagonist with log Kd values of -8 16 -9 08 and -6 93 for 1 2 and 3 respectively