Learn More
Medchemexpress LLC (S)-(-)-Propranolol hydrochloride | 4199-10-4 | 99.9% | 100 MG
SDP
Catalog No. 5000407909
Encompass_Preferred
(S)-(-)-Propranolol hydrochloride is a chiral antihypertensive agent with approximately 100-fold higher potency than its R-enantiomer. It is used for studies on hypertension, angina pectoris, and arrhythmia. It is for research use only and not sold to patients.
  • Chiral antihypertensive agent
  • Approximately 100-fold higher potency than R-enantiomer
  • Used for studies on hypertension
  • Used for studies on angina pectoris
  • Used for studies on arrhythmia
  • For research use only
Catalog No. 50-004-07909 Supplier Medchemexpress LLC Supplier No. HYB0573A100MG
(S)-(-)-Propranolol hydrochloride is a chiral antihypertensive agent with approximately 100-fold higher potency than its R-enantiomer. It is used for studies on hypertension, angina pectoris, and arrhythmia. It is for research use only and not sold to patients.
  • Chiral antihypertensive agent
  • Approximately 100-fold higher potency than R-enantiomer
  • Used for studies on hypertension
  • Used for studies on angina pectoris
  • Used for studies on arrhythmia
  • For research use only