Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-(-)-Propylene Oxide

Catalog No. 54002125G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
54-002-125G 25 g
54-002-15G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 54-002-125G Supplier Sigma Aldrich Supplier No. 54002125G
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(S)-(-)-Propylene oxide undergoes ring-opening upon treating with nucleophiles such as Grignard reagents or organolithium compounds to yield corresponding secondary alcohols.It can be used in the synthesis of ligands to prepare of Pb(II) based coordination nets for the fabrication of white light-emitting diode materials.It is a key starting material for the total synthesis of arenolide and (+)-aspicilin.Manufactured under license by Sterling Pharma Solutions Limited, using Jacobsen HKR technology.

Specifications

CAS 16088-62-3
Density 0.829 g/mL at 20°C
Boiling Point 33°C to 34°C
Molecular Formula C3H6O
Linear Formula C3H6O
Refractive Index n20/D 1.366
MDL Number MFCD00064312
Quantity 25 g
Synonym (S)-(-)-1,2-Epoxypropane; (S)-(-)-Methyloxirane
Molecular Weight (g/mol) 58.08 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.