Learn More
Bon Opus Biosciences Recombinant S Protein RBD-SD1(Mammalian, W436R, C-6His) -500ug
SDP
Supplier:  Bon Opus Biosciences DRA52500UG
Encompass_Preferred
Recombinant 2019-nCoV S protein RBD Protein is produced by our Mammalian expression system and the target gene encoding Arg319-Ser591(W436R) is expressed with a 6His tag at the C-terminus. Expression system: Human Cells. Accession# QHD43416.1. Formulation: Supplied as a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# DRA52
Catalog No. 50-208-0547