Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S,R)-GSK321 | 1816272-18-0 | 98.6% | 10 MG
SDP

Catalog No. 5000338207
Encompass_Preferred

(S,R)-GSK321 is a potent and selective mutant IDH1 inhibitor, specifically the (S,R)-enantiomer of GSK321. It demonstrates high selectivity over IDH2 and induces a decrease in intracellular 2-HG. This compound abrogates the myeloid differentiation block and promotes granulocytic differentiation in leukemic blasts and immature stem-like cells. It is suitable for research into acute myeloid leukemia (AML) and other cancers.

  • Potent, selective mutant IDH1 inhibitor
  • IC50 values of 2.9, 3.8, 4.6 and 46 nM for R132G, R132C, R132H and WT IDH1, respectively
  • Greater than 100-fold selectivity over IDH2
  • Induces a decrease in intracellular 2-HG
  • Abrogates the myeloid differentiation block
  • Induces granulocytic differentiation in leukemic blasts and immature stem-like cells
  • Used for research of acute myeloid leukemia (AML) and other cancers

Catalog No. 50-003-38207 Supplier Medchemexpress LLC Supplier No. HY128888B10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S,R)-GSK321 is a potent and selective mutant IDH1 inhibitor, specifically the (S,R)-enantiomer of GSK321. It demonstrates high selectivity over IDH2 and induces a decrease in intracellular 2-HG. This compound abrogates the myeloid differentiation block and promotes granulocytic differentiation in leukemic blasts and immature stem-like cells. It is suitable for research into acute myeloid leukemia (AML) and other cancers.

  • Potent, selective mutant IDH1 inhibitor
  • IC50 values of 2.9, 3.8, 4.6 and 46 nM for R132G, R132C, R132H and WT IDH1, respectively
  • Greater than 100-fold selectivity over IDH2
  • Induces a decrease in intracellular 2-HG
  • Abrogates the myeloid differentiation block
  • Induces granulocytic differentiation in leukemic blasts and immature stem-like cells
  • Used for research of acute myeloid leukemia (AML) and other cancers

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.