Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S,R)-GSK321 | 1816272-18-0 | 98.6% | 501.55 g/mol | C28H28FN5O3 | 5 MG
SDP

Catalog No. 5000262447
Encompass_Preferred

(S,R)-GSK321 is the (S,R)-enantiomer of GSK321, a potent and selective mutant isocitrate dehydrogenase 1 (IDH1) inhibitor used as a research tool to study IDH1-driven oncogenic metabolism and differentiation. It reduces intracellular 2-hydroxyglutarate (2-HG) levels and promotes granulocytic differentiation in leukemic cells, supporting studies in acute myeloid leukemia and related cancers.

  • Potent mutant IDH1 inhibitor with nanomolar activity.
  • Reduces intracellular 2-hydroxyglutarate (2-HG) levels.
  • Promotes granulocytic differentiation in leukemic cells.
  • High purity suitable for research applications (98.6%).
  • Molecular weight 501.55 g/mol; molecular formula C28H28FN5O3.
  • Available in small research pack sizes, including 5 mg.

Catalog No. 50-002-62447 Supplier Medchemexpress LLC Supplier No. HY128888B5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S,R)-GSK321 is the (S,R)-enantiomer of GSK321, a potent and selective mutant isocitrate dehydrogenase 1 (IDH1) inhibitor used as a research tool to study IDH1-driven oncogenic metabolism and differentiation. It reduces intracellular 2-hydroxyglutarate (2-HG) levels and promotes granulocytic differentiation in leukemic cells, supporting studies in acute myeloid leukemia and related cancers.

  • Potent mutant IDH1 inhibitor with nanomolar activity.
  • Reduces intracellular 2-hydroxyglutarate (2-HG) levels.
  • Promotes granulocytic differentiation in leukemic cells.
  • High purity suitable for research applications (98.6%).
  • Molecular weight 501.55 g/mol; molecular formula C28H28FN5O3.
  • Available in small research pack sizes, including 5 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.