Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S,R,S)-AHPC-amido-C7-acid | 2172819-76-8 | 98.7% | 600.77 | 100 MG
SDP

Catalog No. 5000360046
Encompass_Preferred

(S,R,S)-AHPC-amido-C7-acid incorporates a VHL ligand for the E3 ubiquitin ligase and a PROTAC linker, making it suitable for designing PROTACs. It is for research use only.

  • Incorporates a VHL ligand for E3 ubiquitin ligase
  • Includes a PROTAC linker
  • Suitable for designing PROTACs
  • Purity of 98.71%
  • Molecular weight of 600.77
  • Appears as a solid
  • Color is white to off-white
  • Relevant to cancer research and targeted therapy

Catalog No. 50-003-60046 Supplier Medchemexpress LLC Supplier No. HY148671100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S,R,S)-AHPC-amido-C7-acid incorporates a VHL ligand for the E3 ubiquitin ligase and a PROTAC linker, making it suitable for designing PROTACs. It is for research use only.

  • Incorporates a VHL ligand for E3 ubiquitin ligase
  • Includes a PROTAC linker
  • Suitable for designing PROTACs
  • Purity of 98.71%
  • Molecular weight of 600.77
  • Appears as a solid
  • Color is white to off-white
  • Relevant to cancer research and targeted therapy

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.