Learn More
Medchemexpress LLC (S,R,S)-AHPC-amido-C7-acid | 2172819-76-8 | 98.7% | 600.77 | 25 MG
SDP
Catalog No. 5000363301
Encompass_Preferred
(S,R,S)-AHPC-amido-C7-acid incorporates a VHL ligand for the E3 ubiquitin ligase and a PROTAC linker, making it suitable for PROTAC development and research applications.
  • Incorporates a VHL ligand and a PROTAC linker
  • Suitable for research applications
  • Designed for PROTAC development
Catalog No. 50-003-63301 Supplier Medchemexpress LLC Supplier No. 25MGHY148671
(S,R,S)-AHPC-amido-C7-acid incorporates a VHL ligand for the E3 ubiquitin ligase and a PROTAC linker, making it suitable for PROTAC development and research applications.
  • Incorporates a VHL ligand and a PROTAC linker
  • Suitable for research applications
  • Designed for PROTAC development