Learn More
Medchemexpress LLC (S,R,S)-AHPC-amido-C7-acid | 2172819-76-8 | 98.71% | 600.77 | 500 MG
SDP
Catalog No. 5000361395
Encompass_Preferred
(S,R,S)-AHPC-amido-C7-acid incorporates a VHL ligand for the E3 ubiquitin ligase and a PROTAC linker. It can be used to design PROTACs. This product is for research use only.
  • Incorporates a VHL ligand for E3 ubiquitin ligase
  • Includes a PROTAC linker
  • Used to design PROTACs
  • Relevant for cancer and cancer targeted therapy research
  • Classified as E3 ligase ligand-linker conjugates
Catalog No. 50-003-61395 Supplier Medchemexpress LLC Supplier No. HY148671500MG
(S,R,S)-AHPC-amido-C7-acid incorporates a VHL ligand for the E3 ubiquitin ligase and a PROTAC linker. It can be used to design PROTACs. This product is for research use only.
  • Incorporates a VHL ligand for E3 ubiquitin ligase
  • Includes a PROTAC linker
  • Used to design PROTACs
  • Relevant for cancer and cancer targeted therapy research
  • Classified as E3 ligase ligand-linker conjugates