Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S,R,S)-AHPC-Boc-trans-3-aminocyclobutanol-Pip-CH2COOH | 2086301-47-3 | 99.3% | 754.98 g/mol | C39H58N6O7S | 5 MG
SDP

Catalog No. 5000262798
Encompass_Preferred

(S,R,S)-AHPC-Boc-trans-3-aminocyclobutanol-Pip-CH2COOH is a Boc-protected VHL ligand-linker conjugate used as a building block in proteolysis-targeting chimera (PROTAC) design, providing a VHL-binding moiety and a trans-3-aminocyclobutanol linker for conjugation.

  • Molecular weight 754.98 g/mol.
  • CAS number 2086301-47-3.
  • High purity ≈99.3% (LCMS).
  • Appearance white to off-white solid.
  • Storage -20°C, sealed; in solvent: -80°C (6 months), -20°C (1 month).
  • Solubility DMSO 100 mg/mL; in vivo formulations soluble ≥2.5-5.5 mg/mL.
  • Available pack sizes 1 mg, 5 mg, 10 mg.
  • Application VHL ligand for PROTAC E3 ligase recruitment.

Catalog No. 50-002-62798 Supplier Medchemexpress LLC Supplier No. HY1311685MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S,R,S)-AHPC-Boc-trans-3-aminocyclobutanol-Pip-CH2COOH is a Boc-protected VHL ligand-linker conjugate used as a building block in proteolysis-targeting chimera (PROTAC) design, providing a VHL-binding moiety and a trans-3-aminocyclobutanol linker for conjugation.

  • Molecular weight 754.98 g/mol.
  • CAS number 2086301-47-3.
  • High purity ≈99.3% (LCMS).
  • Appearance white to off-white solid.
  • Storage -20°C, sealed; in solvent: -80°C (6 months), -20°C (1 month).
  • Solubility DMSO 100 mg/mL; in vivo formulations soluble ≥2.5-5.5 mg/mL.
  • Available pack sizes 1 mg, 5 mg, 10 mg.
  • Application VHL ligand for PROTAC E3 ligase recruitment.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.