Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S,R,S)-AHPC-Boc (VH032-Boc) | 1448189-98-7 | MFCD31692982 | 96.4% | 530.68 | C27H38N4O5S | 50MG
SDP

Catalog No. 5000238391
Encompass_Preferred

(S,R,S)-AHPC-Boc (VH032-Boc) is a small-molecule ligand that recruits the von Hippel-Lindau (VHL) E3 ligase and is used as a VHL-recruiting building block in PROTAC design and chemical biology research. The material is supplied as a solid for research use and is intended for synthesis of targeted protein degradation probes and related ligand development.

  • Recruits von Hippel-Lindau (VHL) E3 ligase for PROTACs.
  • Molecular formula C27H38N4O5S; molecular weight 530.68.
  • Purity 96.4%.
  • Available in multiple sizes for synthesis flexibility (10 mg-1 g).
  • Storage: solid at -20°C under nitrogen; in solvent -80°C for 6 months or -20°C for 1 month.
  • Intended for research use in targeted protein degradation and ligand development.

Catalog No. 50-002-38391 Supplier Medchemexpress LLC Supplier No. HY12310950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S,R,S)-AHPC-Boc (VH032-Boc) is a small-molecule ligand that recruits the von Hippel-Lindau (VHL) E3 ligase and is used as a VHL-recruiting building block in PROTAC design and chemical biology research. The material is supplied as a solid for research use and is intended for synthesis of targeted protein degradation probes and related ligand development.

  • Recruits von Hippel-Lindau (VHL) E3 ligase for PROTACs.
  • Molecular formula C27H38N4O5S; molecular weight 530.68.
  • Purity 96.4%.
  • Available in multiple sizes for synthesis flexibility (10 mg-1 g).
  • Storage: solid at -20°C under nitrogen; in solvent -80°C for 6 months or -20°C for 1 month.
  • Intended for research use in targeted protein degradation and ligand development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.