Learn More
Medchemexpress LLC (S,R,S)-AHPC-C10-NHBoc | 2412055-13-9 | 713.97 | 100 MG
SDP
Catalog No. 5000362285
Encompass_Preferred
(S,R,S)-AHPC-C10-NHBoc is a synthesized E3 ligase ligand-linker conjugate that incorporates the (S,R,S)-AHPC based VHL ligand and a linker used for BET-Targeted PROTAC. This conjugate targets E3 ligase ligand-linker conjugates and is involved in the PROTAC pathway.
  • Store powder at -20°C, sealed, away from moisture and light.
  • For solutions, store at -80°C for 6 months or -20°C for 1 month, sealed and away from moisture and light.
Catalog No. 50-003-62285 Supplier Medchemexpress LLC Supplier No. HY148406100MG
Add to Cart
Add to Cart
(S,R,S)-AHPC-C10-NHBoc is a synthesized E3 ligase ligand-linker conjugate that incorporates the (S,R,S)-AHPC based VHL ligand and a linker used for BET-Targeted PROTAC. This conjugate targets E3 ligase ligand-linker conjugates and is involved in the PROTAC pathway.
  • Store powder at -20°C, sealed, away from moisture and light.
  • For solutions, store at -80°C for 6 months or -20°C for 1 month, sealed and away from moisture and light.