Learn More
Medchemexpress LLC (S,R,S)-AHPC-C10-NHBoc | 2412055-13-9 | 713.97 | 50 MG
SDP
Catalog No. 5000362286
Encompass_Preferred
(S,R,S)-AHPC-C10-NHBoc is a synthesized E3 ligase ligand-linker conjugate. It incorporates the (S,R,S)-AHPC based VHL ligand and a linker used for BET-Targeted PROTAC.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates (S,R,S)-AHPC based VHL ligand
  • Used with a linker for BET-targeted PROTAC
Catalog No. 50-003-62286 Supplier Medchemexpress LLC Supplier No. HY14840650MG
(S,R,S)-AHPC-C10-NHBoc is a synthesized E3 ligase ligand-linker conjugate. It incorporates the (S,R,S)-AHPC based VHL ligand and a linker used for BET-Targeted PROTAC.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates (S,R,S)-AHPC based VHL ligand
  • Used with a linker for BET-targeted PROTAC