Learn More
Medchemexpress LLC (S,R,S)-AHPC-C10-NHBoc | 2412055-13-9 | 713.97 g·mol⁻¹ | C38H59N5O6S | 10 MG
SDP
Catalog No. 5000287128
Encompass_Preferred
(S,R,S)-AHPC-C10-NHBoc is a synthesized E3 ligase ligand-linker conjugate that couples the (S,R,S)-AHPC VHL ligand to a 10-carbon alkyl linker. It is designed for use as the VHL-recruiting element in BET-targeted PROTAC design and for studies involving E3 ligase ligand conjugation.
  • Provides a VHL-based E3 ligase recruiting moiety for PROTAC development.
  • Contains a 10-carbon alkyl linker to provide spacer flexibility between ligand and target binder.
  • Designed for chemical synthesis and linker conjugation chemistry.
  • Characterized by molecular formula C38H59N5O6S and molar mass 713.97 g·mol⁻¹.
  • Store sealed away from moisture and light; in solvent: -80°C up to 6 months, -20°C up to 1 month.
Catalog No. 50-002-87128 Supplier Medchemexpress LLC Supplier No. HY14840610MG
(S,R,S)-AHPC-C10-NHBoc is a synthesized E3 ligase ligand-linker conjugate that couples the (S,R,S)-AHPC VHL ligand to a 10-carbon alkyl linker. It is designed for use as the VHL-recruiting element in BET-targeted PROTAC design and for studies involving E3 ligase ligand conjugation.
  • Provides a VHL-based E3 ligase recruiting moiety for PROTAC development.
  • Contains a 10-carbon alkyl linker to provide spacer flexibility between ligand and target binder.
  • Designed for chemical synthesis and linker conjugation chemistry.
  • Characterized by molecular formula C38H59N5O6S and molar mass 713.97 g·mol⁻¹.
  • Store sealed away from moisture and light; in solvent: -80°C up to 6 months, -20°C up to 1 month.