Learn More
Medchemexpress LLC (S,R,S)-AHPC-C10-NHBoc | 2412055-13-9 | 98.3% | 713.97 | 25 MG
SDP
Catalog No. 5000362656
Encompass_Preferred
(S,R,S)-AHPC-C10-NHBoc is a synthesized E3 ligase ligand-linker conjugate. It incorporates the (S,R,S)-AHPC based VHL ligand and a linker, used for BET-Targeted PROTAC technology. This product is intended for laboratory research and the manufacture of substances.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates (S,R,S)-AHPC based VHL ligand
  • Utilized in BET-Targeted PROTAC technology
  • For research use only, not fully validated for medical applications
Catalog No. 50-003-62656 Supplier Medchemexpress LLC Supplier No. 25MGHY148406
(S,R,S)-AHPC-C10-NHBoc is a synthesized E3 ligase ligand-linker conjugate. It incorporates the (S,R,S)-AHPC based VHL ligand and a linker, used for BET-Targeted PROTAC technology. This product is intended for laboratory research and the manufacture of substances.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates (S,R,S)-AHPC based VHL ligand
  • Utilized in BET-Targeted PROTAC technology
  • For research use only, not fully validated for medical applications