Learn More
Medchemexpress LLC (S,R,S)-AHPC-C10-NHBoc | 2412055-13-9 | 98.3% | 713.97 | 5 MG
SDP
Catalog No. 5000361310
Encompass_Preferred
(S,R,S)-AHPC-C10-NHBoc is a synthesized E3 ligase ligand-linker conjugate. It incorporates an (S,R,S)-AHPC based VHL ligand and a linker, designed for BET-Targeted PROTAC research and applications. This product is a laboratory chemical intended for research use only and has not been fully validated for medical applications.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates VHL ligand and a linker
  • Used for BET-targeted PROTAC research
  • Intended for research use only
Catalog No. 50-003-61310 Supplier Medchemexpress LLC Supplier No. HY1484065MG
Add to Cart
Add to Cart
(S,R,S)-AHPC-C10-NHBoc is a synthesized E3 ligase ligand-linker conjugate. It incorporates an (S,R,S)-AHPC based VHL ligand and a linker, designed for BET-Targeted PROTAC research and applications. This product is a laboratory chemical intended for research use only and has not been fully validated for medical applications.
  • Synthesized E3 ligase ligand-linker conjugate
  • Incorporates VHL ligand and a linker
  • Used for BET-targeted PROTAC research
  • Intended for research use only