Learn More
Medchemexpress LLC (S,R,S)-AHPC-C2-amide-benzofuranylmethyl-pyridine | 2347517-69-3 | 98.6% | C41H46N6O6S | 10MG
SDP
Catalog No. 5000259567
Encompass_Preferred
(S,R,S)-AHPC-C2-amide-benzofuranylmethyl-pyridine is a PROTAC small-molecule research reagent that induces ubiquitination and degradation of Smad3 while modulating HIF-α protein levels. It is intended for preclinical studies of anti-fibrosis, renal protection, and oncology mechanisms and is supplied as a high-purity solid.
  • High purity (98.6%) for reliable experimental results.
  • Dual-target PROTAC mechanism induces Smad3 degradation and modulates HIF-α levels.
  • Suitable for preclinical research in fibrosis, renal protection, and oncology.
  • Supplied as a solid and available in small research pack sizes for screening and synthesis.
  • High solubility in DMSO (≥170 mg/mL) for convenient stock preparation.
  • Stable under recommended storage conditions to preserve activity.
Catalog No. 50-002-59567 Supplier Medchemexpress LLC Supplier No. HY14702510MG
(S,R,S)-AHPC-C2-amide-benzofuranylmethyl-pyridine is a PROTAC small-molecule research reagent that induces ubiquitination and degradation of Smad3 while modulating HIF-α protein levels. It is intended for preclinical studies of anti-fibrosis, renal protection, and oncology mechanisms and is supplied as a high-purity solid.
  • High purity (98.6%) for reliable experimental results.
  • Dual-target PROTAC mechanism induces Smad3 degradation and modulates HIF-α levels.
  • Suitable for preclinical research in fibrosis, renal protection, and oncology.
  • Supplied as a solid and available in small research pack sizes for screening and synthesis.
  • High solubility in DMSO (≥170 mg/mL) for convenient stock preparation.
  • Stable under recommended storage conditions to preserve activity.