Learn More
Medchemexpress LLC (S,R,S)-AHPC-C2-amide-benzofuranylmethyl-pyridine | 2347517-69-3 | MFCD34676915 | 98.6% | 750.91 | 1 MG
SDP
Catalog No. 5000360770
Encompass_Preferred
(S,R,S)-AHPC-C2-amide-benzofuranylmethyl-pyridine is a dual-target PROTAC that functions by facilitating the ubiquitination and degradation of Smad3, and by increasing HIF-α protein levels. This compound exhibits multi-path anti-fibrosis activity and renal protection functions.
  • Suitable for research on renal anemia, prostate cancer, and other cancers
  • Facilitates ubiquitination and degradation of Smad3
  • Increases HIF-α protein levels
  • Exhibits multi-path anti-fibrosis function
  • Provides renal protection
Catalog No. 50-003-60770 Supplier Medchemexpress LLC Supplier No. HY1470251MG
(S,R,S)-AHPC-C2-amide-benzofuranylmethyl-pyridine is a dual-target PROTAC that functions by facilitating the ubiquitination and degradation of Smad3, and by increasing HIF-α protein levels. This compound exhibits multi-path anti-fibrosis activity and renal protection functions.
  • Suitable for research on renal anemia, prostate cancer, and other cancers
  • Facilitates ubiquitination and degradation of Smad3
  • Increases HIF-α protein levels
  • Exhibits multi-path anti-fibrosis function
  • Provides renal protection