Learn More
Medchemexpress LLC (S,R,S)-AHPC-C2-PEG3-BCN | 2876198-36-4 | 98.4% | 810.01 | 25 MG
SDP
Catalog No. 5000363938
Encompass_Preferred
(S,R,S)-AHPC-C2-PEG3-BCN is a VHL ligand used for the synthesis of PROTACs. It is intended for research use only and not for patient treatment.
  • VHL ligand
  • Used for synthesis of PROTACs
  • Relevant for cancer and cancer targeted therapy research
  • PROTAC E3 ligase ligand-linker conjugate
Catalog No. 50-003-63938 Supplier Medchemexpress LLC Supplier No. 25MGHY148896
(S,R,S)-AHPC-C2-PEG3-BCN is a VHL ligand used for the synthesis of PROTACs. It is intended for research use only and not for patient treatment.
  • VHL ligand
  • Used for synthesis of PROTACs
  • Relevant for cancer and cancer targeted therapy research
  • PROTAC E3 ligase ligand-linker conjugate