Learn More
Medchemexpress LLC (S,R,S)-AHPC-C2-PEG3-BCN | 2876198-36-4 | 98.4% | 810.01 | 5 MG
SDP
Catalog No. 5000360871
Encompass_Preferred
(S,R,S)-AHPC-C2-PEG3-BCN (Compound 16b) is described as a VHL ligand that can be utilized for the synthesis of PROTACs. It is intended for research use only.
  • VHL ligand
  • Utilized for the synthesis of PROTACs
  • White to light yellow solid
Catalog No. 50-003-60871 Supplier Medchemexpress LLC Supplier No. HY1488965MG
(S,R,S)-AHPC-C2-PEG3-BCN (Compound 16b) is described as a VHL ligand that can be utilized for the synthesis of PROTACs. It is intended for research use only.
  • VHL ligand
  • Utilized for the synthesis of PROTACs
  • White to light yellow solid