Learn More
Medchemexpress LLC (S,R,S)-AHPC-C2-PEG3-BCN | 2876198-36-4 | 98.4% | 810.01 | 50 MG
SDP
Catalog No. 5000359501
Encompass_Preferred
(S,R,S)-AHPC-C2-PEG3-BCN (Compound 16b) is a VHL ligand. It is used for the synthesis of PROTACs.
  • Utilized for the synthesis of PROTACs
  • VHL ligand
  • Related to cancer targeted therapy
  • Inhibitor
Catalog No. 50-003-59501 Supplier Medchemexpress LLC Supplier No. HY14889650MG
(S,R,S)-AHPC-C2-PEG3-BCN (Compound 16b) is a VHL ligand. It is used for the synthesis of PROTACs.
  • Utilized for the synthesis of PROTACs
  • VHL ligand
  • Related to cancer targeted therapy
  • Inhibitor