Learn More
Medchemexpress LLC (S,R,S)-AHPC-C2-PEG3-BCN | 2876198-36-4 | 99.2% | 810.01 g·mol⁻¹ | C42H59N5O9S | 10 MG
SDP
Catalog No. 5000285581
Encompass_Preferred
(S,R,S)-AHPC-C2-PEG3-BCN (Compound 16b) is a VHL E3 ligase ligand-linker conjugate used as a building block in the synthesis of PROTACs and other conjugates. It features a PEG3 spacer and a bicyclononyne (BCN) functional group to facilitate bioorthogonal conjugation and assembly of bifunctional molecules for targeted protein degradation research.
  • VHL E3 ligase ligand for targeted protein degradation applications.
  • PEG3 linker provides a flexible spacer for molecular conjugation.
  • BCN functional group enables strain-promoted bioorthogonal click conjugation.
  • Reported high purity suitable for synthetic chemistry use.
  • Intended as a small-molecule building block for PROTAC assembly and conjugation workflows.
Catalog No. 50-002-85581 Supplier Medchemexpress LLC Supplier No. HY14889610MG
(S,R,S)-AHPC-C2-PEG3-BCN (Compound 16b) is a VHL E3 ligase ligand-linker conjugate used as a building block in the synthesis of PROTACs and other conjugates. It features a PEG3 spacer and a bicyclononyne (BCN) functional group to facilitate bioorthogonal conjugation and assembly of bifunctional molecules for targeted protein degradation research.
  • VHL E3 ligase ligand for targeted protein degradation applications.
  • PEG3 linker provides a flexible spacer for molecular conjugation.
  • BCN functional group enables strain-promoted bioorthogonal click conjugation.
  • Reported high purity suitable for synthetic chemistry use.
  • Intended as a small-molecule building block for PROTAC assembly and conjugation workflows.