Learn More
Medchemexpress LLC (S,R,S)-AHPC-C3-COOH | 2172819-73-5 | C27H36N4O6S | 100 MG
SDP
Catalog No. 5000353446
Encompass_Preferred
(S,R,S)-AHPC-C3-COOH is a carboxylic acid derivative of the VHL E3 ubiquitin ligase ligand-Linker conjugate. This compound can be used to synthesize PROTACs.
  • Acts as a reference compound
  • Targets E3 ligase ligand-linker conjugates
Catalog No. 50-003-53446 Supplier Medchemexpress LLC Supplier No. HY141651100MG
(S,R,S)-AHPC-C3-COOH is a carboxylic acid derivative of the VHL E3 ubiquitin ligase ligand-Linker conjugate. This compound can be used to synthesize PROTACs.
  • Acts as a reference compound
  • Targets E3 ligase ligand-linker conjugates