Learn More
Medchemexpress LLC (S,R,S)-AHPC-(C3-PEG)2-C6-Cl | 1835705-61-7 | MFCD31560481 | 99.9% | 749.44 g/mol | C39H61ClN4O6S | 1 G
SDP
Catalog No. 5000244982
Encompass_Preferred
(S,R,S)-AHPC-(C3-PEG)2-C6-Cl is a HaloPROTAC that couples the (S,R,S)-AHPC VHL ligand to a two-unit PEG linker and a C6-chloroalkyl HaloTag reactive group. It is used in cell-based assays to recruit the von Hippel-Lindau (VHL) E3 ligase and promote degradation of HaloTag fusion proteins.
  • VHL ligand with PEG linker for E3 ligase recruitment.
  • Induces degradation of HaloTag fusion proteins in cell assays.
  • High purity for research applications (99.9%).
  • Solid form, supplied as a 1 g laboratory package.
  • Store at -80°C, protect from light, and store under inert gas.
Catalog No. 50-002-44982 Supplier Medchemexpress LLC Supplier No. HY1036081G
(S,R,S)-AHPC-(C3-PEG)2-C6-Cl is a HaloPROTAC that couples the (S,R,S)-AHPC VHL ligand to a two-unit PEG linker and a C6-chloroalkyl HaloTag reactive group. It is used in cell-based assays to recruit the von Hippel-Lindau (VHL) E3 ligase and promote degradation of HaloTag fusion proteins.
  • VHL ligand with PEG linker for E3 ligase recruitment.
  • Induces degradation of HaloTag fusion proteins in cell assays.
  • High purity for research applications (99.9%).
  • Solid form, supplied as a 1 g laboratory package.
  • Store at -80°C, protect from light, and store under inert gas.