Learn More
Medchemexpress LLC (S,R,S)-AHPC-C4-COOH | 2172819-74-6 | 95.9% | 558.69 | 250 MG
SDP
Catalog No. 5000366086
Encompass_Preferred
(S,R,S)-AHPC-C4-COOH (linker 45) is a synthesized E3 ligase ligand-linker conjugate that incorporates the (S,R,S)-AHPC based VHL ligand and a linker used for EED-targeted PROTAC.
  • Protacs contain two different ligands connected by a linker.
  • One ligand is for an E3 ubiquitin ligase and the other is for the target protein.
  • Protacs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
Catalog No. 50-003-66086 Supplier Medchemexpress LLC Supplier No. 250MGHY135045
(S,R,S)-AHPC-C4-COOH (linker 45) is a synthesized E3 ligase ligand-linker conjugate that incorporates the (S,R,S)-AHPC based VHL ligand and a linker used for EED-targeted PROTAC.
  • Protacs contain two different ligands connected by a linker.
  • One ligand is for an E3 ubiquitin ligase and the other is for the target protein.
  • Protacs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.