Learn More
Medchemexpress LLC HY-112847A 10mg Medchemexpress, Sulfosuccinimidyl oleate (sodium) CAS: Purity:>98%
SDP
Catalog No. 501969190
Encompass_Preferred
Medchemexpress, HY-114176 500mg (S,R,S)-AHPC-C4-NH2 (HCl) CAS:2245697-83-8 (S,R,S)-AHPC-C4-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that incorporates the (S,R,S)-AHPC based VHL ligand and a linker used for EED-Targeted PROTAC. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9190 Supplier Medchemexpress LLC Supplier No. HY112847A10MG
Medchemexpress, HY-114176 500mg (S,R,S)-AHPC-C4-NH2 (HCl) CAS:2245697-83-8 (S,R,S)-AHPC-C4-NH2 hydrochloride is a synthesized E3 ligase ligand-linker conjugate that incorporates the (S,R,S)-AHPC based VHL ligand and a linker used for EED-Targeted PROTAC. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.