Learn More
Medchemexpress LLC (S,R,S)-AHPC-C5-COOH (VH032-C5-COOH) | 2267282-19-7 | >98.0% | 558.74 | C29H42N4O5S | 5 MG
SDP
Catalog No. 5000222405
Encompass_Preferred
(S,R,S)-AHPC-C5-COOH (VH032-C5-COOH) is a synthesized E3 ligase ligand-linker conjugate used as a VHL ligand building block in PROTAC and targeted protein degradation research. It is supplied as a high-purity solid intended for research and medicinal chemistry applications, and is compatible with standard conjugation chemistries.
  • High purity suitable for research applications.
  • Designed as a VHL ligand-linker conjugate for PROTAC synthesis.
  • Supplied as a stable solid with recommended cold storage.
  • Soluble in common organic solvents used for conjugation reactions.
  • Useful as a building block for targeted protein degradation studies.
Catalog No. 50-002-22405 Supplier Medchemexpress LLC Supplier No. HY1360555MG
(S,R,S)-AHPC-C5-COOH (VH032-C5-COOH) is a synthesized E3 ligase ligand-linker conjugate used as a VHL ligand building block in PROTAC and targeted protein degradation research. It is supplied as a high-purity solid intended for research and medicinal chemistry applications, and is compatible with standard conjugation chemistries.
  • High purity suitable for research applications.
  • Designed as a VHL ligand-linker conjugate for PROTAC synthesis.
  • Supplied as a stable solid with recommended cold storage.
  • Soluble in common organic solvents used for conjugation reactions.
  • Useful as a building block for targeted protein degradation studies.