Learn More
Medchemexpress LLC (S,R,S)-ahpc-c5-cooh (vh032-c5-cooh) | 2267282-19-7 | >98.0% | C29H42N4O5S | 10MG
SDP
Catalog No. 5000259767
Encompass_Preferred
(S,R,S)-AHPC-C5-COOH is a von Hippel-Lindau (VHL) E3 ligase ligand-linker conjugate used as a synthetic building block in PROTAC and degrader synthesis. It contains an AHPC recognition motif with a five-carbon linker terminating in a carboxylic acid, enabling facile amide or ester coupling to target ligands. Supplied as a high-purity solid for medicinal chemistry research.
  • High purity (>98%) suitable for synthesis and analytical work.
  • Five-carbon linker terminating in a carboxylic acid for coupling chemistry.
  • Designed for use as a VHL E3 ligase ligand in PROTAC assembly.
  • Provided in small research quantities for medicinal chemistry workflows.
  • Stable solid, typically white to off-white, for convenient handling.
Catalog No. 50-002-59767 Supplier Medchemexpress LLC Supplier No. HY13605510MG
(S,R,S)-AHPC-C5-COOH is a von Hippel-Lindau (VHL) E3 ligase ligand-linker conjugate used as a synthetic building block in PROTAC and degrader synthesis. It contains an AHPC recognition motif with a five-carbon linker terminating in a carboxylic acid, enabling facile amide or ester coupling to target ligands. Supplied as a high-purity solid for medicinal chemistry research.
  • High purity (>98%) suitable for synthesis and analytical work.
  • Five-carbon linker terminating in a carboxylic acid for coupling chemistry.
  • Designed for use as a VHL E3 ligase ligand in PROTAC assembly.
  • Provided in small research quantities for medicinal chemistry workflows.
  • Stable solid, typically white to off-white, for convenient handling.