Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S,R,S)-AHPC-C5-NH2 dihydrochloride | 2415256-20-9 | 97.8% | 616.64 | 25 MG
SDP

Catalog No. 5000347841
Encompass_Preferred

(S,R,S)-AHPC-C5-NH2 dihydrochloride is a chemical compound with a molecular weight of 616.64. It appears as an off-white to light yellow solid and is suitable for research applications. This product is for research use only and has not been fully validated for medical applications.

  • Chemical name: (2S,4R)-1-((S)-2-(6-Aminohexanamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide dihydrochloride
  • CAS number: 2415256-20-9
  • Molecular formula: C28H43Cl2N5O4S
  • Molecular weight: 616.64
  • Purity: 97.8% (HPLC)
  • Appearance: Off-white to light yellow solid
  • Storage: -20°C, sealed storage, away from moisture. In solvent: -80°C for 6 months; -20°C for 1 month, sealed storage, away from moisture.

Catalog No. 50-003-47841 Supplier Medchemexpress LLC Supplier No. 25MGHY136187A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S,R,S)-AHPC-C5-NH2 dihydrochloride is a chemical compound with a molecular weight of 616.64. It appears as an off-white to light yellow solid and is suitable for research applications. This product is for research use only and has not been fully validated for medical applications.

  • Chemical name: (2S,4R)-1-((S)-2-(6-Aminohexanamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-(4-(4-methylthiazol-5-yl)benzyl)pyrrolidine-2-carboxamide dihydrochloride
  • CAS number: 2415256-20-9
  • Molecular formula: C28H43Cl2N5O4S
  • Molecular weight: 616.64
  • Purity: 97.8% (HPLC)
  • Appearance: Off-white to light yellow solid
  • Storage: -20°C, sealed storage, away from moisture. In solvent: -80°C for 6 months; -20°C for 1 month, sealed storage, away from moisture.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.