Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S,R,S)-AHPC-C5-NH2 dihydrochloride | 2415256-20-9 | 97.8% | 616.64 g/mol | C28H43Cl2N5O4S | 5 MG
SDP

Catalog No. 5000222243
Encompass_Preferred

(S,R,S)-AHPC-C5-NH2 dihydrochloride is the dihydrochloride salt of an AHPC-derived E3 ligase ligand, provided as a solid for research use in studies of VHL E3 ubiquitin ligase recruitment and targeted protein degradation.

  • Dihydrochloride salt form for improved stability.
  • High purity of 97.8%.
  • Molecular weight 616.64 g/mol.
  • Molecular formula C28H43Cl2N5O4S.
  • Suitable as an E3 ligase ligand-linker component in PROTAC designs.
  • Storage: sealed, away from moisture; in solvent: -80°C up to 6 months, -20°C up to 1 month.
  • Documentation available: datasheet, COA, SDS.

Catalog No. 50-002-22243 Supplier Medchemexpress LLC Supplier No. HY136187A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S,R,S)-AHPC-C5-NH2 dihydrochloride is the dihydrochloride salt of an AHPC-derived E3 ligase ligand, provided as a solid for research use in studies of VHL E3 ubiquitin ligase recruitment and targeted protein degradation.

  • Dihydrochloride salt form for improved stability.
  • High purity of 97.8%.
  • Molecular weight 616.64 g/mol.
  • Molecular formula C28H43Cl2N5O4S.
  • Suitable as an E3 ligase ligand-linker component in PROTAC designs.
  • Storage: sealed, away from moisture; in solvent: -80°C up to 6 months, -20°C up to 1 month.
  • Documentation available: datasheet, COA, SDS.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.