Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S,R,S)-AHPC-C5-NH2 dihydrochloride | 2415256-20-9 | 97.8% | C28H43Cl2N5O4S | 10MG
SDP

Catalog No. 5000261163
Encompass_Preferred

(S,R,S)-AHPC-C5-NH2 dihydrochloride is a VHL ligand-linker conjugate supplied as the dihydrochloride salt for use as a PROTAC building block in research. It contains a C5 linker terminating in a primary amine and is provided as a solid intended for laboratory synthesis and biological studies.

  • Purity 97.8%.
  • Off-white to light yellow solid appearance.
  • Chemical formula C28H43Cl2N5O4S.
  • Molecular weight 616.64 g/mol.
  • Recommended storage: -20°C, sealed and away from moisture.
  • In solvent stability: -80°C for 6 months; -20°C for 1 month.
  • For research use only; not for human use.

Catalog No. 50-002-61163 Supplier Medchemexpress LLC Supplier No. HY136187A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S,R,S)-AHPC-C5-NH2 dihydrochloride is a VHL ligand-linker conjugate supplied as the dihydrochloride salt for use as a PROTAC building block in research. It contains a C5 linker terminating in a primary amine and is provided as a solid intended for laboratory synthesis and biological studies.

  • Purity 97.8%.
  • Off-white to light yellow solid appearance.
  • Chemical formula C28H43Cl2N5O4S.
  • Molecular weight 616.64 g/mol.
  • Recommended storage: -20°C, sealed and away from moisture.
  • In solvent stability: -80°C for 6 months; -20°C for 1 month.
  • For research use only; not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.