Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S,R,S)-Ahpc-C6-Peg3-C4-Cl | 1835705-55-9 | MFCD31560478 | ≥95.0% | 751.42 g/mol | C38H59ClN4O7S | 500 MG
SDP

Catalog No. 5000247794
Encompass_Preferred

(S,R,S)-AHPC-C6-PEG3-C4-Cl is a VHL ligand-linker conjugate used as a building block in targeted protein degradation (PROTAC) research. It supplies a halide-terminated PEG linker designed for coupling in degrader synthesis and related medicinal chemistry applications.

  • Used as a VHL ligand-linker building block for PROTAC synthesis.
  • Molecular formula C38H59ClN4O7S.
  • Molecular weight 751.42 g/mol.
  • Purity ≥95.0% (manufacturer assay).
  • Supplied as a 500 mg solid.
  • Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-47794 Supplier Medchemexpress LLC Supplier No. HY103605500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S,R,S)-AHPC-C6-PEG3-C4-Cl is a VHL ligand-linker conjugate used as a building block in targeted protein degradation (PROTAC) research. It supplies a halide-terminated PEG linker designed for coupling in degrader synthesis and related medicinal chemistry applications.

  • Used as a VHL ligand-linker building block for PROTAC synthesis.
  • Molecular formula C38H59ClN4O7S.
  • Molecular weight 751.42 g/mol.
  • Purity ≥95.0% (manufacturer assay).
  • Supplied as a 500 mg solid.
  • Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.