Learn More
Medchemexpress LLC (S,R,S)-AHPC-C6-PEG3-C4-Cl | 1835705-55-9 | MFCD31560478 | 97.0% | 751.42 | C38H59ClN4O7S | 1 G
SDP
Catalog No. 5000245153
Encompass_Preferred
(S,R,S)-AHPC-C6-PEG3-C4-Cl is a VHL ligand-linker conjugate designed as a building block for PROTAC synthesis and ligand-linker conjugate development in targeted protein degradation research. It contains an (S,R,S)-configured AHPC VHL ligand connected via a six-carbon PEG3 linker with a terminal chloro group to enable downstream conjugation.
  • Useful as a VHL (E3 ligase) ligand building block for degrader design.
  • C6-PEG3 linker provides flexibility and spacer length for bifunctional molecules.
  • Terminal chloro group enables nucleophilic substitution or further functionalization.
  • Characterized by analytical data including HPLC and LC-MS for batch confirmation.
  • Provided as a solid suitable for synthetic chemistry workflows and storage under controlled conditions.
Catalog No. 50-002-45153 Supplier Medchemexpress LLC Supplier No. HY1036051G
(S,R,S)-AHPC-C6-PEG3-C4-Cl is a VHL ligand-linker conjugate designed as a building block for PROTAC synthesis and ligand-linker conjugate development in targeted protein degradation research. It contains an (S,R,S)-configured AHPC VHL ligand connected via a six-carbon PEG3 linker with a terminal chloro group to enable downstream conjugation.
  • Useful as a VHL (E3 ligase) ligand building block for degrader design.
  • C6-PEG3 linker provides flexibility and spacer length for bifunctional molecules.
  • Terminal chloro group enables nucleophilic substitution or further functionalization.
  • Characterized by analytical data including HPLC and LC-MS for batch confirmation.
  • Provided as a solid suitable for synthetic chemistry workflows and storage under controlled conditions.