Learn More
Medchemexpress LLC HY-101763A 1g Medchemexpress, (S,R,S)-AHPC (hydrochloride) CAS:1448189-80-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY101763A1G
Encompass_Preferred
Medchemexpress, HY-101763A 1g (S,R,S)-AHPC (hydrochloride) CAS:1448189-80-7 (S,R,S)-AHPC hydrochloride (MDK7526 hydrochloride) is the VH032-based VHL ligand used in the recruitment of the von Hippel-Lindau (VHL) protein. (S,R,S)-AHPC hydrochloride can be connected to the ligand for protein (e.g., BCR-ABL1) by a linker to form PROTACs (e.g., GMB-475). GMB-475 induces the degradation of BCR-ABL1 with an IC 50 of 1.11 μM in Ba/F3 cells [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2207