Learn More
Medchemexpress LLC HY-101763A 500mg Medchemexpress, (S,R,S)-AHPC (hydrochloride) CAS:1448189-80-7 Purity:>98%
SDP
Catalog No. 502028675
Encompass_Preferred
Medchemexpress, HY-101763A 500mg (S,R,S)-AHPC (hydrochloride) CAS:1448189-80-7 (S,R,S)-AHPC hydrochloride is the VH032-based VHL ligand used in the recruitment of the von Hippel-Lindau (VHL) protein. (S,R,S)-AHPC hydrochloride can be connected to the ligand for protein (e.g., BCR-ABL1) by a linker to form PROTACs (e.g., GMB-475). GMB-475 induces the degradation of BCR-ABL1 with an IC50 of 1.11 μM in Ba/F3 cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8675 Supplier Medchemexpress LLC Supplier No. HY101763A500MG
Medchemexpress, HY-101763A 500mg (S,R,S)-AHPC (hydrochloride) CAS:1448189-80-7 (S,R,S)-AHPC hydrochloride is the VH032-based VHL ligand used in the recruitment of the von Hippel-Lindau (VHL) protein. (S,R,S)-AHPC hydrochloride can be connected to the ligand for protein (e.g., BCR-ABL1) by a linker to form PROTACs (e.g., GMB-475). GMB-475 induces the degradation of BCR-ABL1 with an IC50 of 1.11 μM in Ba/F3 cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.