Learn More
Selleck Chemical LLC (S.R.S)-AHPC hydrochloride S1062-1g
SDP
Catalog No. 503108419
Encompass_Preferred
(S R S)-AHPC hydrochloride (VH032-NH2 VHL ligand 1) is the VH032-based VHL ligand that is applicable to the recruitment of the von Hippel-Lindau (VHL) protein (S R S)-AHPC hydrochloride can be connected to the ligand for protein by a linker to form PROTACs
Catalog No. 50-310-8419 Supplier Selleck Chemical LLC Supplier No. S10621G
(S R S)-AHPC hydrochloride (VH032-NH2 VHL ligand 1) is the VH032-based VHL ligand that is applicable to the recruitment of the von Hippel-Lindau (VHL) protein (S R S)-AHPC hydrochloride can be connected to the ligand for protein by a linker to form PROTACs