Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S,R,S)-AHPC-Me-C10-Br | 2836297-55-1 | 98.0% | C34H51BrN4O4S | 10MG
SDP

Catalog No. 5000260511
Encompass_Preferred

(S,R,S)-AHPC-Me-C10-Br is a synthesized E3 ligase ligand-linker conjugate designed for targeted protein degradation research. The compound links a VHL E3 ligase ligand to an MS432-derived warhead and is supplied for research use only.

  • E3 ligase ligand-linker conjugate for proteolysis-targeting chimera (PROTAC) development.
  • Incorporates a VHL E3 ligase linker.
  • Contains an MS432-derived warhead based on a MEK1/2 inhibitor.
  • High purity suitable for research applications.
  • Molecular formula C34H51BrN4O4S and molecular weight 691.76.
  • Supplied as a 10 mg quantity.

Catalog No. 50-002-60511 Supplier Medchemexpress LLC Supplier No. HY13064210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S,R,S)-AHPC-Me-C10-Br is a synthesized E3 ligase ligand-linker conjugate designed for targeted protein degradation research. The compound links a VHL E3 ligase ligand to an MS432-derived warhead and is supplied for research use only.

  • E3 ligase ligand-linker conjugate for proteolysis-targeting chimera (PROTAC) development.
  • Incorporates a VHL E3 ligase linker.
  • Contains an MS432-derived warhead based on a MEK1/2 inhibitor.
  • High purity suitable for research applications.
  • Molecular formula C34H51BrN4O4S and molecular weight 691.76.
  • Supplied as a 10 mg quantity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.