Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S,R,S)-AHPC-Me-C7 ester | 2639528-48-4 | 95.49% | 100 MG
SDP

Catalog No. 5000338166
Encompass_Preferred

(S,R,S)-AHPC-Me-C7 ester is an E3 ligase ligand-linker conjugate utilized for synthesizing BCL-XL PROTAC degraders. This product is for research use only.

  • Molecular weight: 614.80
  • Formula: C32H46N4O6S
  • Appearance: Solid
  • Color: White to off-white
  • Solubility: 6.67 mg/mL (10.85 mM) in DMSO
  • Shipping: Room temperature in continental US
  • Storage: -20°C, sealed, away from moisture; in solvent: -80°C for 6 months or -20°C for 1 month

Catalog No. 50-003-38166 Supplier Medchemexpress LLC Supplier No. HY130640100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S,R,S)-AHPC-Me-C7 ester is an E3 ligase ligand-linker conjugate utilized for synthesizing BCL-XL PROTAC degraders. This product is for research use only.

  • Molecular weight: 614.80
  • Formula: C32H46N4O6S
  • Appearance: Solid
  • Color: White to off-white
  • Solubility: 6.67 mg/mL (10.85 mM) in DMSO
  • Shipping: Room temperature in continental US
  • Storage: -20°C, sealed, away from moisture; in solvent: -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.