Learn More
Medchemexpress LLC HY-42424A 100mg Medchemexpress, (S,R,S)-AHPC-Me (dihydrochloride) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY42424A100MG
Encompass_Preferred
Medchemexpress, HY-42424A 100mg (S,R,S)-AHPC-Me (dihydrochloride) CAS: (S,R,S)-AHPC-Me dihydrochloride (VHL ligand 2 dihydrochloride) is the (S,R,S)-AHPC-based VHL ligand used in the recruitment of the von Hippel-Lindau (VHL) protein [1] . (S,R,S)-AHPC-Me dihydrochloride can be used to synthesize ARV-771, a von Hippel-Landau (VHL) E3 ligase-based BET PROTAC degrader. ARV-771 potently degrades BET protein in castration-resistant prostate cancer (CRPC) cells with a DC 50 <1 nM [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2860