Learn More
Medchemexpress LLC (S,R,S)-AHPC-Me-PEG2-dabrafenib | 2769753-69-5 | 99.9% | 1106.31 | 25 MG
SDP
Catalog No. 5000350553
Encompass_Preferred
DD-03-156 is a potent and selective degrader of CDK17 and LIMK2. Its selectivity and potency make it an advanced starting point for developing a chemical probe for the degradation of CDK17.
  • Potent and selective degrader of CDK17 and LIMK2
  • Advanced starting point for developing a chemical probe for CDK17 degradation
  • Purity of 99.9%
  • White to off-white solid appearance
  • Consistent with structure by 1H NMR and LCMS
Catalog No. 50-003-50553 Supplier Medchemexpress LLC Supplier No. 25MGHY137346
DD-03-156 is a potent and selective degrader of CDK17 and LIMK2. Its selectivity and potency make it an advanced starting point for developing a chemical probe for the degradation of CDK17.
  • Potent and selective degrader of CDK17 and LIMK2
  • Advanced starting point for developing a chemical probe for CDK17 degradation
  • Purity of 99.9%
  • White to off-white solid appearance
  • Consistent with structure by 1H NMR and LCMS