Learn More
Medchemexpress LLC (S,R,S)-AHPC-Me-PEG2-dabrafenib | 2769753-69-5 | 99.9% | 1106.31 g/mol | C53H62F3N9O8S3 | 5 MG
SDP
Catalog No. 5000221849
Encompass_Preferred
DD-03-156 ((S,R,S)-AHPC-Me-PEG2-dabrafenib) is a selective small-molecule degrader developed for research use that targets cyclin-dependent kinase 17 (CDK17) and LIM kinase 2 (LIMK2). The compound is provided as a high-purity solid in milligram-scale quantities and is intended for biochemical and cell-based studies with recommended low-temperature storage under inert atmosphere.
  • Selective degrader of CDK17 and LIMK2.
  • High reported purity suitable for research assays.
  • Supplied in milligram-scale quantities for exploratory studies.
  • Stable under recommended low-temperature storage when handled under inert atmosphere.
  • Includes molecular characterization and CAS registry number for traceability.
Catalog No. 50-002-21849 Supplier Medchemexpress LLC Supplier No. HY1373465MG
DD-03-156 ((S,R,S)-AHPC-Me-PEG2-dabrafenib) is a selective small-molecule degrader developed for research use that targets cyclin-dependent kinase 17 (CDK17) and LIM kinase 2 (LIMK2). The compound is provided as a high-purity solid in milligram-scale quantities and is intended for biochemical and cell-based studies with recommended low-temperature storage under inert atmosphere.
  • Selective degrader of CDK17 and LIMK2.
  • High reported purity suitable for research assays.
  • Supplied in milligram-scale quantities for exploratory studies.
  • Stable under recommended low-temperature storage when handled under inert atmosphere.
  • Includes molecular characterization and CAS registry number for traceability.